Learn More
Kartell™ Plastilab™ L-Shaped Untapered Connectors
Click to view available options
Port Size:
4.5mm bore
6 mm
6mm bore
8 mm bore
8mm bore
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
Description
- Made from polypropylene
- Autoclavable
- Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
| Packaging Quantity | 10/Pk. |
| Port Size | 6 mm |
| Shape | L-shaped |
| Product Type | Untapered L Tubing Connector |