Learn More
Kartell™ Plastilab™ L-Shaped Untapered Connectors
BIOPHARMA
Product Code. p-8000643
Click to view available options
Port Size:
4.5mm bore
6 mm
6mm bore
8 mm bore
8mm bore
Packaging Quantity:
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
Product Code. 12539766
Brand: Kartell™ 7125200
Only null left
Add to basket
Edge
Add to basket
Designed for use with hose system and vacuum lines.
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Certifications/Compliance Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Diameter (Metric) 8 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Useful for any hoses system and vacuum line
Packaging Quantity 10/Pk.
Port Size 4.5mm bore
Shape L-shaped
Product Type Untapered L Tubing Connector