Learn More
Smeg™ Upper Trolley Rack
Clean a wide range of glassware with this basic trolley rack. Smeg™ Upper Trolley Racks are designed for use with Smeg glassware washers.
Brand: Smeg™ CS1-1
Additional Details : Weight : 2.40000kg
Description
- Upper rack with spray arm CS1-1
- Made for stainless steel and suitable for positioning supports
- Dimensions: 468 x 481 x 151mm
Specifications
Upper Trolley Rack | |
For positioning Stainless Steel baskets and supports | |
For Smeg 60cm trolley washer | |
1 Each |