missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Liofilchem™ Urea/Indole Broth
Click to view available options
Quantity:
100 mL
Unit Size:
Pack of 6
Description
- Medium for urease and indole test.
Specifications
Specifications
| Content And Storage | 10°C to 25°C |
| Manufacturing Origin | Italy |
| Product Type | Microbiology Media |
| Sterility | Non-sterile |
| Target Organism Class | Gram-negative Bacteria, Coliforms |
| Quantity | 100 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction