Learn More
Alla France™ Urine Hydrometer
Product Code. 11708093
Click to view available options
Length (Metric):
165 mm
Unit Size:
Each
Product Code. 11708093
Brand: Alla France™ 1080PM060/20qp
Only null left
Add to basket
Add to basket
Order Info
dimLengthMetric
Specifications
Quantity 1Each
Type Urine Hydrometer
Length (Metric) 165 mm
Specific Gravity Range 1.000 to 1.060 g/mL