missing translation for 'onlineSavingsMsg'
Learn More

Biochrom UV Grade Silica Micro Cell with Lid and Black Walls

10 mm Quartz Cell with lid and 2 black walls. 10 mm pathlength with 400 μL minimum volume.

Brand:  Biochrom 80-2002-95

Additional Details : Weight : 0.02800kg

Product Code. 11300142

  • 615.00€ / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • 10 mm Quartz Cell with lid and 2 black walls
  • 10 mm pathlength
  • Minimum volume: 400 μL
  • Fits standard spectrophotometer cell holders
  • For use with Ultrospec 7000/8000/9000, Ultrospec 2100 Pro, GeneQuant 1300/100
Specifications

Specifications

Type 8 Micro Cuvette
UV Grade Silica
1 Ea.
Dual Beam Ultrospec Spectrophotometers
400 μL
2 mm
10 mm
Spectrophotometric Measurements
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions