Learn More
UVP UV Viewing Cabinets
Click to view available options
Model:
C-65
C-70G
Unit Size:
Each
Description
- Soft rubber viewport includes a contrast control filter.
- Control switches are conveniently located for selecting either of the UV wavelengths or the interior overhead white light
- Easy access to the cabinet interior is via the curtain which also blocks ambient light
- Manufactured from lightweight plastic
- The C-65 cabinet can hold one or two EL Series 8-watt lamps (sold separately)
Specifications
Specifications
| Voltage | 230V |
| Depth (Metric) | 409 mm |
| Height (Metric) | 400 mm |
| Width (Metric) | 493 mm |
| Model | C-65 |
| Packaging Quantity | 1Pack |
| For Use With (Application) | Thin Layer Chromatography, Non Destructive Testing, Inspection, Mineralogy, Fluorescence |
| Product Type | UV Chamber |