Learn More
TCR V alpha 2 Monoclonal Antibody (B20.1), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
119.00€ - 333.00€
Specifications
Antigen | TCR V alpha 2 |
---|---|
Clone | B20.1 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15350760
|
Affymetrix eBioscience
17-5812-80 |
25 μg |
119.00€
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15360760
|
Affymetrix eBioscience
17-5812-82 |
100 μg |
333.00€
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The B20.1 monoclonal antibody reacts with mouse T-cell receptor (TCR) V alpha 2 chains. This antibody recognizes the majority of the TCR V alpha 2 subfamily in mice carrying the a, b and c haplotypes. The antibody also reacts with the products of T cell receptor, V delta 8 due to the high degree of homology. Applications Reported: This B20.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This B20.1 antibody has been tested by flow cytometric analysis of mouse lymph node cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.It is recommended to preblock Fc receptors for 10 minutes using 0.5 μg of purified anti-mouse CD16/32 (cat. 14-0161). It is also recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
TCR V alpha 2 Monoclonal antibody specifically detects TCR V alpha 2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V alpha 2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a λ | |
Affinity chromatography | |
Antibody |
B20.1 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
TCRVA; TCRVA2; V alpha 2; V alpha-2 TCR; Va 2; Va2; Valpha 2; Valpha2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.