Learn More
Wheaton™ Vaccum Filter with Frit
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
574.00€ - 805.00€
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
10498711
|
Wheaton™
419520-47 |
47 mm |
574.00€
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
|||||
11751883
|
Wheaton™
419452 |
90 mm |
805.00€
Each |
Estimated Shipment: 07-07-2025
Log in to see stock. |
|||||
Description
Wheaton Vaccum Filter with Frit, For Use With: To Collect The Filtrate Directly Into A Standard Laboratory Bottle
Specifications
Vaccum Filter |