missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ PET SST II Advance Tubes

Product Code. 12567876
Click to view available options
Material:
PET
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12567876

Brand: BD 366882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Product Type Advance Tube
Material PET
Sterility Sterile
Closure Type Hemogard
Capacity (Metric) 2.5 mL
Closure Color Gold
Tube Size 13 x 75 mm
Includes Pk of 100
For Use With (Application) Clinical evaluation of special chemistries such as therapeutic drugs, proteins, peptides, steroids and vitamins
Label Type Block
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.