Learn More
- Home
- Products
- Ovens and Furnaces
- Oven Accessories
- Thermo Scientific™ Vacutherm™ Oven Accessories and Options
Thermo Scientific™ Vacutherm™ Oven Accessories and Options
Product Code. 10408631 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Aluminum tray
Central Monitoring Connection
Digicon Temperature Controller
Digital Pressure Display
Digital Timer
Door
Door Gasket
Precision Control Valve
Pressure Controller
Shelf
Stainless-steel tray
Support Frame
Vacuum Chamber
Vacuum Connection Kit
Weekly program timer, digital
For Use With (Equipment):
VT 6025
VT 6025/6060/6130 M/P
VT 6060 M
VT 6060 M/P and VT6130 M/P
VT 6060 P
VT 6060 P,VT 6130 P
VT 6130 M
VT 6130 M/P
VT 6130 P
VT6060 M/P
Unit Size:
1 set
Each
25 product options available for selection
| Product Code. | Type | For Use With (Equipment) | unitSize |
|---|---|---|---|
| 10408631 | Vacuum Connection Kit | VT 6025/6060/6130 M/P | 1 set |
| 10383192 | Pressure Controller | VT 6060 M/P and VT6130 M/P | Each |
| 11671661 | Digicon Temperature Controller | VT 6060 P,VT 6130 P | Each |
| 10482014 | Digicon Temperature Controller | VT 6060 P | Each |
| 10213924 | Digicon Temperature Controller | VT 6130 P | Each |
| 10517611 | Digital Timer | VT 6060 M/P and VT6130 M/P | Each |
| 10321002 | Weekly program timer, digital | VT 6060 M/P and VT6130 M/P | Each |
| 10472614 | Door | VT 6130 P | Each |
| 10397603 | Door | VT 6060 P | Each |
| 10376434 | Vacuum Chamber | VT 6130 P | Each |
| 11611631 | Vacuum Chamber | VT 6060 M | Each |
| 11621631 | Vacuum Chamber | VT 6130 M | Each |
| 10490884 | Vacuum Chamber | VT 6060 P | Each |
| 11661621 | Digital Pressure Display | VT 6060 M/P and VT6130 M/P | Each |
| 10100991 | Door Gasket | VT 6060 M | Each |
| 11681621 | Door Gasket | VT 6130 M | Each |
| 11671631 | Central Monitoring Connection | VT 6060 M/P and VT6130 M/P | Each |
| 11567199 | Precision Control Valve | VT 6025 | Each |
| 11696440 | Shelf | VT 6025 | Each |
| 11691471 | Shelf | VT 6130 M | Each |
| 10467281 | Support Frame | VT 6130 M/P | Each |
| 12778696 | Support Frame | VT6060 M/P | Each |
| 11621521 | Stainless-steel tray | VT 6130 M/P | Each |
| 10337442 | Stainless-steel tray | VT6060 M/P | Each |
| 10520875 | Aluminum tray | VT6060 M/P | Each |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
Description
Choose from a range of Thermo Scientific™ Vacutherm™ Oven Accessories and Options to meet application-specific requirements.
Specifications
Specifications
| Description | Stainless-steel vacuum connection kit for tubing, 10mm diamenter |
| For Use With (Equipment) | VT 6025/6060/6130 M/P |
| Type | Vacuum Connection Kit |
| Material | Stainless Steel |