Learn More
BRANDTECH™ VACUUBRAND™ Electronic Vacuum Gauges
GREENER_CHOICE
Product Code. p-7146487
Click to view available options
Maximum Pressure:
0.001 to 1100 mbar
0.1 to 1100 mbar
1 to 1060 mbar
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 15838894
Brand: BRANDTECH™ VACUUBRAND™ 20683220
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
VACUUBRAND vacuum gauges are mercury-free. Choose from models for the coarse/rough vacuum range or fine vacuum range. User-selectable units.
  • DVR2pro model is battery-powered for portability, with a capacitive transducer for absolute pressure measurements from atmosphere to 1 mbar/Torr/hPA. Combination digital/analog scale for easy reading.
  • VACUU VIEW model features a capacitive transducer for absolute pressure measurement and measures to 0.1 mbar/Torr/hPA. Universal power suppy.
  • VACUU VIEW extended, has a combination capacitive/Pirani transducer for measurements to 10-3 mbar/Torr/hPA. Universal power supply.
TRUSTED_SUSTAINABILITY
Specifications
Display Digital
For Use With (Application) Vacuum applications in the rough/coarse vacuum range atmosphere - 0.1 mbarTorr/hPa
For Use With Vacuum applications
Includes Vacuum gauge, power supply, KF16 small flange connection and screw in 6/10mm hose barb
Material Various
Product Line VACUU VIEW
Type Capacitive Vacuum Gauge
Weight (Metric) 1 kg
Height (Metric) 8.5 cm
Length (Metric) 25 cm
Width (Metric) 16 cm
Maximum Pressure 0.1 to 1100 mbar