Learn More
Gardner Denver Welch™ Kit 9 Vacuum pump control package with MRV 100, DN 16 KF
Shop All Welch Ilmvac Products
Click to view available options
Unit Size:
Each
Description
Vacuum control package consists of:
- MRV100 vacuum controller
- PIZA111cr-GOLD vacuum sensor
- KF16 measuring T-piece
- 2-way solenoid valve
- Clamps, centring rings and adapters
- Electrical connections
Specifications
Specifications
| Description | DN 16 KF |
| Product Line | Vacuum Control Kit |
| For Use With | KF16 flanged Rotary Vane Pumps |