Learn More
Merck Vacuum Filtering Side-arm Flask
Accessories for filter holders for sample preparation.
Brand: Merck XX1014744
434.06 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.69400kg
Description
Accessories for filter holders for sample preparation.
- Vacuum filtering side-arm flask
- Designed for use in vacuum filtration procedures with classic Millipore Glass Filter Holders
- This is an accessory item for the Glass Microanalysis Filter Holder Kit
Features & Benefits:
- Side arm connects to vacuum source with 3/8 in. I.D. hose
- 4 L flasks accept a No. 8 perforated stopper
- Applications: Vacuum Filtration Using Classic Filter Holders
Note: Stopper is not included; sold separately.
Specifications
Clear | |
Borosilicate Glass | |
Filtering Flask | |
4 L |
Threaded side-arm | |
Vacuum Filtration Using Classic Filter Holders | |
Accepts a No. 8 perforated silicone stopper (not included) |