missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Valentino S3 Shoe

Product Code. p-7225049
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
This item is not returnable. View return policy

Product Code. 17572446

Brand: Emma Safety Footwear MM16400039

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Break new ground in the Valentino Frontier, the executive safety shoe that literally has no boundaries.

  • Industries:
    • Chemical industry
    • Government & services
    • Oil & gas
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work

Specifications

Certifications/Compliance 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Chemical Industry, Government and Services, Oil and Gas
Material Leather
Size 39
Toe Material Steel
Fit Extra-wide Width
Ankle Style High
Product Type Safety Shoe
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.