missing translation for 'onlineSavingsMsg'
Learn More

Vanadium(III) 2,4-pentanedionate, typically 97%, Thermo Scientific Chemicals

Catalog Number p-7049798
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
13476-99-8
C15H21O6V
348.27
MFCD00000033
ZVFSRLXFKIWGRT-UHFFFAOYSA-N
Vanadium(III) acetylacetonate
122198782
(Z)-4-hydroxypent-3-en-2-one;vanadium
[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
This item is not returnable. View return policy
13476-99-8
C15H21O6V
348.27
MFCD00000033
ZVFSRLXFKIWGRT-UHFFFAOYSA-N
Vanadium(III) acetylacetonate
122198782
(Z)-4-hydroxypent-3-en-2-one;vanadium
[V+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Vanadium(III) 2,4-pentanedionate is used as homogeneous catalyst, primary and secondary intermediates, Laboratory chemicals, and manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
UN Number UN3285
Sensitivity Air Sensitive
Solubility Information Soluble in water 2g/L,toluene,ethanol,acetone.
Formula Weight 348.27
Percent Purity 97% (typically)
Physical Form Powder
Chemical Name or Material Vanadium(III) 2,4-pentanedionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'