missing translation for 'onlineSavingsMsg'
Learn More

Vandetanib, TRC

Product Code. 30453602
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
Product Code. Quantity unitSize
30453602 10 mg 10mg
30456708 5 mg 5mg
30457063 100 mg 100mg
3 options
This item is not returnable. View return policy

Product Code. 30453602

Brand: Toronto Research Chemicals TRCV09710010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443913-73-3
Formula Weight 474.1067
InChI Formula InChI=1S/C22H24BrFN4O2/c1-28-7-5-14(6-8-28)12-30-21-11-19-16(10-20(21)29-2)22(26-13-25-19)27-18-4-3-15(23)9-17(18)24/h3-4,9-11,13-14H,5-8,12H2,1-2H3,(H,25,26,27)
IUPAC Name N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
Molecular Formula C22 H24 Br F N4 O2
Molecular Weight (g/mol) 475.35
SMILES COc1cc2c(Nc3ccc(Br)cc3F)ncnc2cc1OCC4CCN(C)CC4
Synonym 4-Quinazolinamine, N-(4-bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine, 4-(4-Bromo-2-fluoroanilino)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazoline, CH 331, Caprelsa, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-(N-methylpiperidin-4-ylmethoxy)quinazolin-4-amine, N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine, Vandetanib, ZD 6474, Zactima
Chemical Name or Material Vandetanib

Specifications

Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.