Learn More
Testo™ Vane Measurement Probe with Telescope
Use the 16 mm diameter probe to determine flow velocity or volume flow in ventilation ducts. It is particularly suitable for medium flow velocities up to + 50 m/s and delivers optimum results.
Brand: Testo™ 0635 9542
Additional Details : Weight : 0.50000kg
Description
- Use the vane/temperature probe (16 mm diameter) to determine flow velocity or volume flow in ventilation ducts (e.g. during the maintenance, evaluation and commissioning of ventilation systems)
- It is particularly suitable for medium flow velocities up to + 50 m/s and delivers optimum results
- The vane anemometer can also measure temperatures up to +70 °C at the same time
- Digital probe connection with integrated calibration concept
- PC software for fast analysis, archiving and documentation of measurement and customer data
- High-quality digital probes with testo's intelligent calibration concept
- Standards-compliant HVAC and comfort level measurements
Specifications
Vane/temperature probe with telescope, Protective cap, and Calibration protocol | |
+118.1 to +9842 fpm / +0.6 to +50 m/s | |
960 mm |
±1.8°C, ±(39.4 fpm + 1% of mv) (118.1 to +7874 fpm) / ±(0.2 m/s + 1% of mv) (0.6 to +40 m/s) | |
-10°C to +70°C |