Learn More
Cole-Parmer™ VapLock™ Activated Media Exhaust Filters
VapLock™ Activated Media Exhaust Filters reduce harmful chemical emissions and odors from solvent containers
94.00€ - 125.00€
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | VapLock Solvent Containment Systems |
Thread Size (English) | 1/4 in. NPT(F) |
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Material | Price | Quantity & Availability | ||||
17225093
|
Cole-Parmer™
12019-79 |
Exhaust Filter, 75 g activated carbon, 1/4 in. NPT(F); 1/ea | Activated Carbon |
94.00€
Each |
In Stock
Log in to see stock. |
||||
16682033
|
Cole-Parmer™
12019-80 |
Exhaust Filter with Indicator, 75 g activated carbon, 1/4 in. NPT(F); 1/ea | Activated Carbon with Breakthrough Indicator |
125.00€
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
||||
Description
Use with indicators to set a replacement scheduleWarranty
None
Specifications
Solvent Safety | |
VapLock Solvent Containment Systems | |
1/4 in. NPT(F) |