Learn More
Macherey-Nagel™ VarioPrep™ Nucleodur™ C18 Isis HPLC Columns
Shop All Macherey-Nagel Products
Click to view available options
Length (Metric):
50 mm
125 mm
250 mm
Carbon Load:
17.5%
18%
20%
Unit Size:
Each
Specifications
Specifications
| Column Format | Preparative Column |
| Diameter (Metric) | 10 mm |
| For Use With | Steroids, (o,p,m-) Substituted Aromatics, Fat-Soluble Vitamins |
| Length (Metric) | 125 mm |
| Max. Pressure | 5800 psi (400 bar) |
| Particle Size | 5 μm |
| pH | 1 to 10 |
| Pore Size | 110 Å |
| Surface Area | 340 m²/g |
| Temperature | 60°C |
| Show More |