Learn More
Emma Safety Footwear Venus XD S3 Safety Shoes
The Venus is a low profile, easy on / easy off shoe. The smooth, easy to clean contours offer additional hygiene advantages over and above lace up shoes. It's perfect for a 'quick change' from office to factory floor or walking between one facility and another.
Brand:  Emma Safety Footwear MM79456646
Additional Details : Weight : 1.79700kg
 View more versions of this product
Product Code. 15979051
  • 125.08€ / 1 pair
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • Full grain leather construction
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Specifications
Venus XD S3
Leather
46