Learn More
Veolia Water Systems™ UV Lamp
Product Code. p-8014593
Click to view available options
For Use With (Equipment):
Bactericide and to break down and photo-oxidize organic contaminants to polar or ionized species
PURELAB Option-R
Unit Size:
Each
Product Code. 11954897
Brand: Veolia Water Systems™ LC105
Only null left
Add to basket
Add to basket
Rich, robust UV lamp that features UV light acting as a bactericide to break down and photo-oxidize organic contaminants to polar or ionized species.
  • UV light is used as a bactericide and to break down and photo-oxidise organic contaminants to polar or ionized species
  • The UV bactericidal action damages DNA and RNA polymerase at low doses thereby preventing replication
Specifications
For Use With (Application) Break down and photo-oxidize organic contaminants to polar or ionized species
For Use With (Equipment) PURELAB Option-R