Learn More
Emma Safety Footwear Vera Safety Shoes, Blue
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
Brand:  Emma Safety Footwear MM50651447
Additional Details : Weight : 1.57800kg
 View more versions of this product
Product Code. 15938511
  • 126.14€ / 1 pair
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Duo Outsole
  • Microfiber construction
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
White
Hydro-Tec™ Sanitized Silver
47
Elastic
Vera, Blue
Microfiber
Steel