Learn More
Emma Safety Footwear Vera XD Safety Shoes, Green
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and dirt resistant.
Brand:  Emma Safety Footwear MM50750438
Additional Details : Weight : 1.24000kg
 View more versions of this product
Product Code. 15928571
  • 126.14€ / 1 pair
Estimated Shipment: 21-06-2024
to see stock.
Add to basket
Description
Description
  • S2-Safety shoe for wet working conditions
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD certified
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
Vera XD, Green
Microfiber
38