Learn More
Emma Safety Footwear Vera XD Safety Shoes, Red
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Description
- S2-Safety shoe for wet working conditions
- Microfiber construction
- GripForce™ Duo Heat Resistant Outsole
- Chrome-free lining
- Sanitized
Specifications
Specifications
| Design | Vera XD, Red |
| Material | Microfiber |
| Size | 38 |