Learn More
Emma Safety Footwear Vera XD Safety Shoes, Red
Product Code. 15988591
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. 15988591
Brand: Emma Safety Footwear MM50752438
Only null left
Add to basket
Add to basket
An easy shoe for wipe clean hygiene. The upper is fully washable unique microfiber and features Hydro-Tec™ Sanitized Silver for dry, fresh feet. The protective toe cap prevents staining and is lubricant and fat resistant. Developed for use on hard tiled floors and other easy-to-clean floors.
  • S2-Safety shoe for wet working conditions
  • Microfiber construction
  • GripForce™ Duo Heat Resistant Outsole
  • Chrome-free lining
  • Sanitized
Specifications
Design Vera XD, Red
Material Microfiber
Size 38