Learn More
3M™ Versaflo™ S-Series Headcovers and Hoods
Click to view available options
Product Type:
Hood with Faceseal
Hood with Inner Shroud
Hood with Neck Collar
Painter's Hood with Inner Shroud Assembly
Respirator Hood
Color:
Gray
White
Model:
S-433 L
S-433 L-5
S-433 S
S-605-10
S-607-10
S-655
S-657
S-707-10
S-757
S-805-5
S-855E
Förpackningsstorlek
Each
Pack of 10
Pack of 5
Beskrivelse
- S-Series respiratory headcovers and hoods are specifically designed to work together with your Versaflo™ Powered Air Respirator and 3M™ Supplied Air Respirator Systems.
- Provides head, face, neck and shoulder coverage from dust and paint spray.
- Provides a good field of view.
- Helps protect eyes and face from liquid splash and low energy flying particles and dust.
- Versatile accessories available that can help you adapt the hood to your particular environment.
Tekniske data
Tekniske data
| Description | Premium Hood with Sealed Seam, Single Size, S-855E |
| Color | Gray |
| Material | Spunbound Polypropylene |
| Product Type | Respirator Hood |
| Format | Standard |
| For Use With (Equipment) | 3M Powered and supplied air respirator systems |
| Includes | Head harness and S-805E replacement fabric hood |
| Model | S-855E |
| Quantity | 1 Ea. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion