Learn More
3M™ Versaflo™ TR-600 PAPR Accessory, Powered Air Turbo Unit, TR-602E
Product Code. 15529305
Click to view available options
:
Powered Air Turbo Unit, TR-602E
Product Code. 15529305
Brand: 3M™ 7100035056
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For use with 3M™ Versaflo™ TR-600 series. 3M Versaflo™ TR-600 PAPR Accessory, Powered Air Turbo Unit, TR-602E includes Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2).
  • Includes an Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2)
  • Comfortable Airflow - Three user selectable flow rates to maximise comfort. Controlled airflow delivers steady flow as battery discharges or filter loads with particulates
  • Display shows battery charge status and particulate filter loading status during use
  • Charge status is also indicated by LED display on battery
  • Electronic audible, visual and vibratory alarm in case of low battery and/or low air flow
  • No user calibration of turbo required
  • Two belt options available: Easy Clean Belt - submersible belt with smooth surfaces allows easy, thorough cleaning and High Durability Belt for hot work or rugged environments which provides greater durability and spark/flame resistance
  • Approved to EN 12941 Class TH2 or TH3 dependent on headtop.
  • Inlet plugs for full submersion and cleanup, meets IP54 and IP67 ratings for use in decontamination shower and temporary submersion when used with cleaning and storage plugs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Powered Air Turbo Unit
For Use With (Equipment) 3M Versaflo TR-600 series
Includes Airflow Indicator (TR-971) and Spark Arrestor (TR-662 x2)
Model TR-602E
Weight (Metric) 670 g