Learn More
Ansell™ Versatouch™ 72-286 Series Dyneema™ Fiber Medium Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
Pack of 6
Description
- Washable up to 90°C
- Offers comfort and durability
- Knitted construction
- Polypropylene Glass fiber Dyneema™ liner
- Knitwrist cuff
Specifications
Specifications
| Back Material | Dyneema™ Fiber |
| Certifications/Compliance | CE Mark, Standard CATEGORY II EN 388 : 254X |
| Cuff Material | Dyneema™ Fiber |
| Coating Material | Uncoated |
| Color | Blue |
| Cut Resistance Rating | Level D |
| Hand | Specific |
| Latex Free | Yes |
| Material | Dyneema, Polyester, Spandex |
| Product Type | Mediumweight Gloves |
| Show More |