missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Versatouch™ 72-287 Series Dyneema™ Fiber Heavy Weight Gloves

Product Code. 12619771 Shop All Ansell Products
Click to view available options
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 12619771

Brand: Ansell Edmont™ 72287090

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Excellent cut performance with ergonomic design to improve comfort. Suitable for contact with all foodstuffs.

  • Contains Dyneema™
  • High cut resistance
  • Washable up to 90°C
  • Suitable for contact with all foodstuffs

Applications:

  • Boning
  • Carving
  • Fish processing
  • Fish filleting
  • Meals preparation
  • Fruit and vegetable processing
TRUSTED_SUSTAINABILITY

Specifications

Back Material Dyneema™ Fiber
Certifications/Compliance CE Mark, Standard CATEGORY II EN 388 : 254X
Cuff Material Dyneema™ Fiber
Product Type Heavyweight Gloves
Glove Cuff Style Knitwrist
Liner Material Dyneema™ Fiber
Lining Dyneema™ Fiber
Quantity 72 Pairs
Resistant To Cut resistant
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.