Learn More
Honeywell™ Vertigo Black NIT C&G 1 Gloves
Description
- Black polyamide knitted glove
- Black foam nitrile coating on palm and fingertips
- The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior comfort and ventilation
- With their ergonomical design the gloves conform to the the natural shape of the hand for a more comfortable fit and enable longer use and less fatigue
- The filament yarn reduces linting for the most sensitive applications and can be easily laundered for extended use
- The foam nitrile coating provides superior and excellent grip and abrasion resistance
- The black color makes the glove less dirty
- The open back provides superior ventilation
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards |
| Description | 13 gauge |
| Coating Material | Polyamide, Nitrile |
| Color | Black |
| Size | 10 |
| Length (Metric) | 24.5 cm |