Learn More
Honeywell™ Vertigo Black NIT C&G 1 Gloves
Product Code. 15844095
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
1 pair
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15844095 10 Each
15846715 7 1 pair
15856715 8 1 pair
15866715 9 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15844095 Supplier Honeywell™ Supplier No. 223227010
Only null left
Add to basket
Edge
Add to basket
Honeywell™ Vertigo Black NIT C&G 1 Gloves for handling of fine products in a greasy, damp or dirty atmosphere.
  • Black polyamide knitted glove
  • Black foam nitrile coating on palm and fingertips
  • The ergonomic seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior comfort and ventilation
  • With their ergonomical design the gloves conform to the the natural shape of the hand for a more comfortable fit and enable longer use and less fatigue
  • The filament yarn reduces linting for the most sensitive applications and can be easily laundered for extended use
  • The foam nitrile coating provides superior and excellent grip and abrasion resistance
  • The black color makes the glove less dirty
  • The open back provides superior ventilation
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards
Description 13 gauge
Coating Material Polyamide, Nitrile
Color Black
Size 10
Length (Metric) 24.5 cm