Learn More
RUBIS Full Plastic Tweezers, ESD Safe
Product Code. 11719574
Encompass_Preferred
Click to view available options
Tip Style:
Angled, Pointed, Ultra Fine
Pointed, Ultra Fine
Round, Flat
Squared, Fine
Straight, Round, Flat
Unit Size:
Each
Product Code. 11719574
Brand: RUBIS NK7
Only null left
Add to basket
Add to basket
Full Plastic Tweezers - Vestamid
  • Vestamid Plastic
  • ESD Safe
  • Avoids scratches on delicate components
  • Very good mechanical resistance
  • Easy to use
  • High Precision
  • 100% Made in Switzerland
Specifications
Tip Style Pointed, Ultra Fine
Length (English) 4.72 in.
Material ESD Safe Vestamid Plastic
Length (Metric) 120 mm
Product Type Plastic Tweezers
Quantity (No. per Case) Each
Type Tweezers