Learn More
RUBIS Full Plastic Tweezers, ESD Safe
Click to view available options
Tip Style:
Angled, Pointed, Ultra Fine
Pointed, Ultra Fine
Round, Flat
Squared, Fine
Straight, Round, Flat
Unit Size:
Each
Description
- Vestamid Plastic
- ESD Safe
- Avoids scratches on delicate components
- Very good mechanical resistance
- Easy to use
- High Precision
- 100% Made in Switzerland
Specifications
Specifications
| Tip Style | Angled, Pointed, Ultra Fine |
| Length (English) | 4.72 in. |
| Material | ESD Safe Vestamid Plastic |
| Length (Metric) | 120 mm |
| Product Type | Plastic Tweezers |
| Quantity (No. per Case) | Each |
| Type | Tweezers |