missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 032

Description
VH 032 is an inhibitor of E3 ubiquitin ligase VHL (Kd = 185 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. VH 032 treatment causes dose dependent upregulation of HIF-target genes including CA9, GLUT1 and PHD2 in different cell lines. Cell permeable.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-Acetamido-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide |
| CAS | 1448188-62-2 |
| Quantity | 50 mg |
| Target | Von Hippel Lindau |
| Molecular Formula | C24H32N4O4S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?