missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 032 amide-alkylC10-amine

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
VH 032 amide-alkylC10-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was VH 032 - linker 10
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2S,4R)-1-((S)-2-(11-Aminoundecanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride |
| Synonym | VH 032 - linker 10 |
| Quantity | 25 mg |
| Molecular Formula | C33H51N5O4S.2HCl |
| Formula Weight | 686.78 |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction