missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 032 amide-alkylC7-amine

Description
VH 032 amide-alkylC7-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus an alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-2-(8-Aminooctanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride |
| CAS | 2415256-17-4 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C30H45N5O4S0.2 HCl |
| Formula Weight | 2020 |
| Purity | 95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?