missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VH 032 amide-PEG5-acid

Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
VH 032 amide-PEG5-acid is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG5 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | -21-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidine-1-carbonyl)-22,22-dimethyl-19-oxo-4,7,10,13,16-pentaoxa-20-azatricosanoic acid |
| CAS | 2172820-14-1 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C36H54N4O11S |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction