Learn More
WTW™ Potassium Reaction Cuvette
Product Code. 10639503
missing translation for 'orderingAttributeHoverText'
Type:
Potassium Reaction Cuvette
missing translation for 'unitSize'
Each
Product Code. 10639503
missing translation for 'mfr': WTW™ 252020
Specifications
Type Potassium Reaction Cuvette
For Use With (Application) Drinking Water, Surface Water, Spring and Well Water, Mineral and Curative Water, Nutrient Solutions for Fertilization, Soils
Diameter (Metric) 16 mm
compliance-icons compliance-icons compliance-icons
Product Identifier
  • K 00615 Reaktionsküvetten Kalium, 16mm Durchm., 30 300mg/l K, 25 Bestimmungen
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : mercury(II) sulphate