Learn More
Serva Electrophoresis™ Proteinase K from Tritirachium album
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
Each
Specifications
Specifications
| Concentration | 8 DMC μ/mg |
| Host Species | Tritirachium album |
| For Use With (Application) | 1 DMC-unit catalyzes the hydrolysis of 1 μmole dimethyl casein per minute at 25°C, pH 7.0; the liberated amino groups are determined with 2,4,6-trinitrobenzene sulfonic acid (2) |
| Source | Proteinase K |
| Quantity | 100 mg |
| Product Type | Proteinase K from Tritirachium album |
| Molecular Weight (g/mol) | 28000 |
| Inhibitors | AEBSF (cat. no. 12745), (PEFABLOC™ SC (cat. no. 31682), PMSF (cat. no. 32395) and diisopropylfluorophosphate (cat. no. 77205). |
Safety and Handling
Product Identifier
- Proteinase K from Tritirachium album min. 8 DMC-U/mg
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .