Learn More
Enzo Life Sciences 1mg Ganglioside GD3 . disodium salt (bovine brain)
Brand:  Enzo Life Sciences ALX-302-010-M001
Encompass_Preferred
1mg Ganglioside GD3 . disodium salt (bovine brain)
Product Code. 15108927
  • 525.14€ / 1mg
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ganglioside GD3 . disodium salt (bovine buttermilk)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.