Learn More
Tenak™ Cardboard Rack for Upright Freezers, 50 mm Box Height, with Boxes
Product Code. 15211004 Shop All TENAK Products
Click to view available options
Holds:
Boxes (16)
Boxes (20)
Boxes (24)
Unit Size:
Each
Product Code. 15211004
Brand: Tenak™ TE26136B
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Due to the design and choice of material, these unique racks are extremely solid and long lasting.
  • Gives the end user a second to none opportunity for reducing costs
  • Ideal for long term storage or for applications where price is of high priority
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Height (Metric) 50 mm (Box)
Dimensions (L x W x H) 562 x 139 x 331 mm
For Use With (Equipment) Upright Freezers
Holds Boxes (24)
Array 6 x 4
Material Cardboard