Learn More
DiscoverX ChemiScreen™ β3 Adrenergic Receptor Stable Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The ChemiScreen™ β3 Adrenergic Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRB3 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses ADRB3 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of ADRB3 , the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Calcium Flux |
| Cell Line | Chem-10 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Epinephrine |
| Quantity | 2 Vials |
| Target Entry | ADRB3 |
| Accession Number | NM_000025 |