Learn More
Invitrogen™ Human RFT1 (aa 445-469) Control Fragment Recombinant Protein
Product Code. 30203233
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30203233
Brand: Invitrogen™ RP106999
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-66318 (PA5-66318, PA5-84799 (PA5-84799. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be involved in N-linked oligosaccharide assembly. May participate in the translocation of oligosaccharide from the cytoplasmic side to the lumenal side of the endoplasmic reticulum membrane.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q96AA3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 91869
Name Human RFT1 (aa 445-469) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CDG1N; congenital disorder of glycosylation 1 N; protein RFT1 homolog; putative endoplasmic reticulum multispan transmembrane protein; RFT1; RFT1 homolog; RFT1, requiring fifty three 1 homolog
Common Name RFT1
Gene Symbol RFT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QSLCFIHRYYRRSPHRPLAGLHLSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining