Learn More
Invitrogen™ Human SLC22A23 (aa 619-686) Control Fragment Recombinant Protein
Product Code. 30209381
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30209381
Brand: Invitrogen™ RP91150
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53214 (PA5-53214. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number A1A5C7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 63027
Name Human SLC22A23 (aa 619-686) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 3110004L20Rik; C6orf85; Ion transporter protein; Nritp; Slc22a23; Solute carrier family 22 member 23; solute carrier family 22, member 23
Common Name SLC22A23
Gene Symbol Slc22a23
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PESRDQNLPENISNGEHYTRQPLLPHKKGEQPLLLTNAELKDYSGLHDAAAAGDTLPEGATANGMKAM
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining