missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean 100™ Vinyl Cleanroom Gloves

Product Code. 15208248
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 15208248

Brand: Nitritex™ BVAL

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

BioClean 100™ vinyl cleanroom gloves are ideal for Class 10 environments.

  • Vinyl Cleanroom Gloves are fully compatible with Class 10 operating environments
  • Double-bagged in anti-static bags for critical environment use
  • Washed in de-ionised water and packed under Class 10 conditions
  • Non-sterile, powder-free, ambidextrous gloves that have been independently tested and found to have extremely low levels of particulates
  • Formulated for user comfort, excellent tactility and strength
  • Micro-roughened for improved grip and are resistant to a wide range of chemicals
  • 300mm length provides added user protection in the cuff area
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ISO 9001:2015 and (EU) 2016/425
Classification Category 1
Color Clear
Disposable Single-use
Latex Free Yes
Material Vinyl
Powdered Powder Free
Product Type Cleanroom Gloves
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded/Rolled
Model BVA-LGE
Quantity 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces)
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.