missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl with Polyester Overlaminate Durable Marking Tape

Product Code. 11791615
Click to view available options
Color:
Black, White
Black, Yellow
Magenta, Yellow
Red, White
Dimensions (L x W):
16.5 m x 25 mm
16.5 m x 38 mm
16.5 m x 50 mm
16.5 m x 75 mm
Unit Size:
1 roll
This item is not returnable. View return policy

Product Code. 11791615

Brand: Brady™ 055302

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Dimensions (L x W) 16.5 m x 50 mm
Finish High Gloss
For Use With (Application) Safety marking of obstructions, low hanging objects, pillars, passageways, aisles, and any other potentially hazardous areas or objects
Length (Metric) 16.5 m
Packaging Quantity 1 Roll
Temperature Range (Metric) -40°C to +80°C
Thickness 0.037 mm
Width (Metric) 50 mm
Adhesive Type Rubber-based Pressure-Sensitive
Color Black, Yellow
Material Vinyl
Product Type Durable Marking Tape
Type Marking Tape
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.