missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11189-100
497.25 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| HIV-1 Gag p24 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 155030 | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 1042942 | |
| Western Blot 1 ug/mL, ELISA | |
| Q77YG1_9HIV1 | |
| E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1_9HIV1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction