Learn More
Ansell™ Virtex™ 79-700 Series Blue Nitrile Immersion Gloves
Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
Unit Size:
50 pairs
Description
- Reusable
- Anti-static according to EN1149
- With Aquadri™-Ansell Moisture Management Technology™, which gives excellent moisture absorption capabilities and keeps users' hands drier for longer
- Significantly lower tendency to shed than traditional cotton flock gloves
- Ideal for light assembly in dry, fully wet or oily conditions, engine and system testing, paint and spray shops, food distribution
Specifications
Specifications
| Back Material | Nitrile |
| Cuff Material | Nitrile |
| Product Type | Immersion Gloves |
| Size | 10 |
| Style | 79-700 |
| Thickness (Metric) | 0.2 mm |
| Glove Cuff Color | Blue |
| Glove Cuff Style | Straight |
| Liner Material | Aquadri |
| Quantity | 50 Pairs |
| Show More |