Learn More
Macherey-Nagel™ Visocolor™ HE Ammonium High Sensitive Titration Kit
Brand:  Macherey-Nagel™ 920006
Additional Details : Weight : 1.00000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 10015220
  • 113.00€ / 1 test
Estimated Shipment: 02-08-2024
to see stock.
Add to basket
Specifications
1 Each
0 to 0.50mg/L NH4
110 Determinations
HE Ammonium High Sensitive Titration Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOUR HE ammonium
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, tri-sodium citrate, sodium nitroprusside,sodium saliylate