Learn More
Macherey-Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits
Run a wide range of water testing assays. Macherey Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits avoid hazardous substances wherever possible.
Brand: Macherey-Nagel™ 931010
Additional Details : Weight : 0.19800kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Colorimetric and titrimetric test kits
- Avoids hazardous substances wherever possible
Specifications
To determine ammonia in water | |
40 g | |
50 Tests | |
Visocolor Eco Ammonium 15 |
30mL NH4-1, 2.5 g NH4-2, 6mL NH4-3, 70mm measuring spoon, 2 screw-plug measuring glasses, slide comparator, color chart, 1m plastic syringe, instructions for use | |
0 · 0.5 · 1 · 2 · 3 · 5 · 7 · 10 · 15mg/L NH4+ | |
Colorimetric method |
Safety and Handling
- VISOCOLOR ECO Ammonium 15
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide solution, ethanol, thymol