Learn More
Aalborg™ Viton A O-ring Stainless Steel Frame with Plug
Product Code. p-8008840
Click to view available options
Size:
2-tube, 150 mm
3-tube, 150 mm
4-tube, 150 mm
Unit Size:
Each
Product Code. 11748538
Brand: Aalborg™ P31S2VA0
Only null left
Add to basket
Add to basket
Aalborg™ Viton A O-ring Stainless Steel Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
  • Rib-guided or fluted metering tubes facilitate stable, accurate readings
  • Interchangeability of flow tubes and floats
  • Manifolding at inlet or outlet
  • Ease of installation and exchange of flow tubes
  • Chemical compatibility
  • Simple means of mounting
Specifications
For Use With (Equipment) Gas/Liquid Flowmeter
Type Frame
Includes Plug
Size 3-tube, 150 mm